LIF161662818 Life Chemicals Inc.

2-(Ethoxycarbonyl)-1H-indole-5-carboxylic acid

  • MDL number MFCD09027048
  • Cas Number : 138731-14-3
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart