LIF161662822 Life Chemicals Inc.

3-(2,5-Dimethoxyphenyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD03030181
  • Cas Number : 882238-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart