LIF161662829 Life Chemicals Inc.

1-Ethyl-8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD11058036
  • Cas Number : 1018135-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart