LIF161662858 Life Chemicals Inc.

5-Oxo-2,5-dihydro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD08741874
  • Cas Number : 37832-55-6
  • Molecular Weight : 128.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart