LIF180875177 Life Chemicals Inc.

4-(2-aminophenyl)piperazin-2-one

  • MDL number MFCD09040656
  • Cas Number : 926199-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart