LIF181847607 Life Chemicals Inc.

2-(5-Amino-3-tert-butyl-1H-pyrazol-1-yl)-6-methylpyrimidin-4(3H)-one

  • MDL number MFCD11518879
  • Cas Number : 1170651-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart