LIF181848562 Life Chemicals Inc.

3-Hydrazino-6-methyl-5H-[1,2,4]triazino[5,6-b]indole

  • MDL number MFCD00998270
  • Cas Number : 330446-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart