LIF181848569 Life Chemicals Inc.

4-amino-8-fluoroquinoline-3-carboxylic acid

  • MDL number MFCD10037653
  • Cas Number : 476683-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart