LIF181848571 Life Chemicals Inc.

4-Amino-6-fluoroquinoline-3-carboxylic acid

  • MDL number MFCD11053981
  • Cas Number : 933688-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart