LIF181848575 Life Chemicals Inc.

1-(3-Chloro-2-methylphenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD01461764
  • Cas Number : 63674-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart