LIF181848576 Life Chemicals Inc.

8-Chloro-2-phenylquinoline-4-carboxylic acid

  • MDL number MFCD00487562
  • Cas Number : 181048-56-6
  • Molecular Weight : 283.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart