LIF184887444 Life Chemicals Inc.

(6,7-Dimethyl-5-oxo-2,3-dihydro-5H-[1,3]thiazolo[3,2-a]pyrimidin-3-yl)acetic acid

  • MDL number MFCD11986422
  • Cas Number : 1105190-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart