LIF184887445 Life Chemicals Inc.

(5-Oxo-2,3,6,7,8,9-hexahydro-5H-[1,3]thiazolo[2,3-b]quinazolin-3-yl)acetic acid

  • MDL number MFCD11518878
  • Cas Number : 1105190-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart