LIF184888065 Life Chemicals Inc.

isocyanatocyclopropane

  • MDL number MFCD02093694
  • Cas Number : 4747-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart