LIF184888411 Life Chemicals Inc.

4-(Chloromethyl)-2-phenyl-1,3-thiazole

  • MDL number MFCD00723503
  • Cas Number : 4771-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart