LIF189455652 Life Chemicals Inc.

8-bromo-1,2,3,4-tetrahydro-1,6-naphthyridine

  • MDL number MFCD11847918
  • Cas Number : 362606-16-4
  • Molecular Weight : 213.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart