LIF189455664 Life Chemicals Inc.

2-(2-aminoethyl)-6-(2-furyl)pyridazin-3(2{H})-one

  • MDL number MFCD11984634
  • Cas Number : 1105192-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart