LIF189455744 Life Chemicals Inc.

6-(2,4-dimethyl-1,3-thiazol-5-yl)pyridazin-3-ol

  • MDL number MFCD16631566
  • Cas Number : 1105195-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart