LIF189455812 Life Chemicals Inc.

(5-oxo-2,3,5,6,7,8-hexahydrocyclopenta[d][1,3]thiazolo[3,2-a]pyrimidin-3-yl)acetic acid

  • MDL number MFCD11986423
  • Cas Number : 1125409-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart