LIF189456117 Life Chemicals Inc.

4-{[(2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl)oxy]methyl}benzoic acid

  • MDL number MFCD11986724
  • Cas Number : 1105194-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart