LIF189456153 Life Chemicals Inc.

3-methoxy-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11986419
  • Cas Number : 113100-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart