LIF189456202 Life Chemicals Inc.

5-(2-methylphenyl)isoxazole-3-carboxylic acid

  • MDL number MFCD07377105
  • Cas Number : 668970-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart