LIF189456256 Life Chemicals Inc.

5-amino-1H-pyrazol-3-ol

  • MDL number MFCD00233932
  • Cas Number : 28491-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart