LIF189456351 Life Chemicals Inc.

6-pyridin-2-ylpyridazin-3-ol

  • MDL number MFCD00114939
  • Cas Number : 66317-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart