LIF189456353 Life Chemicals Inc.

3-(3,4,5-trimethoxyphenyl)-1{H}-pyrazole-5-carboxylic acid

  • MDL number MFCD05170217
  • Cas Number : 1319125-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart