LIF189456388 Life Chemicals Inc.

1-methyl-3-(2-thienyl)-1{H}-pyrazole-5-carboxylic acid

  • MDL number MFCD08271954
  • Cas Number : 871825-56-8
  • Molecular Weight : 208.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart