LIF189456409 Life Chemicals Inc.

3-chloro-6-(thiophen-2-yl)pyridazine

  • MDL number MFCD00114988
  • Cas Number : 28657-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart