LIF189456416 Life Chemicals Inc.

3-chloro-6-pyridin-3-ylpyridazine

  • MDL number MFCD11984636
  • Cas Number : 78784-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart