LIF189456422 Life Chemicals Inc.

6-morpholin-4-ylpyridazin-3-ol

  • MDL number MFCD15194890
  • Cas Number : 27464-00-2
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart