LIF189456428 Life Chemicals Inc.

2-(2-Aminoethyl)pyridazin-3(2{H})-one

  • MDL number MFCD11984633
  • Cas Number : 215524-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart