LIF189456431 Life Chemicals Inc.

(6-oxopyridazin-1(6{H})-yl)acetic acid

  • MDL number MFCD03990569
  • Cas Number : 95209-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart