LIF189456473 Life Chemicals Inc.

3-(4-oxo-1,2,3-benzotriazin-3(4H)-yl)propanoic acid

  • MDL number MFCD02926470
  • Cas Number : 51672-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart