LIF189456505 Life Chemicals Inc.

4-chloro-6-(pyridin-3-yl)pyrimidine

  • MDL number MFCD09750196
  • Cas Number : 954232-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart