LIF189456543 Life Chemicals Inc.

3-amino-1,5-dimethyl-1,4,5,7-tetrahydro-6H-pyrazolo[3,4-b]pyridin-6-one

  • MDL number MFCD14279118
  • Cas Number : 871021-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart