LIF189457144 Life Chemicals Inc.

5-Amino-1-ethyl-1H-1,2,3-triazole-4-carboxamide

  • MDL number MFCD12400946
  • Cas Number : 99420-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart