LIF189457214 Life Chemicals Inc.

tert-Butyl 4-(8-amino-5-isoquinolinyl)-1-piperazinecarboxylate

  • MDL number MFCD16631744
  • Cas Number : 1105191-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart