LIF189457235 Life Chemicals Inc.

2-{[4-(methylsulfonyl)phenyl]amino}-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD11986873
  • Cas Number : 1105191-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart