LIF189457268 Life Chemicals Inc.

N-[4-(trifluoromethyl)phenyl]guanidine

  • MDL number MFCD10000732
  • Cas Number : 130066-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart