LIF189457315 Life Chemicals Inc.

2-(pyridin-4-yl)-1-(4-(quinolin-2-ylmethoxy)phenyl)ethan-1-one

  • MDL number MFCD09743764
  • Cas Number : 871507-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart