LIF189457478 Life Chemicals Inc.

5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD05864580
  • Cas Number : 7268-43-1
  • Molecular Weight : 129.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart