LIF189457483 Life Chemicals Inc.

5-(3,4,5-triethoxyphenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09804613
  • Cas Number : 19949-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart