LIF189457517 Life Chemicals Inc.

5-(Chloromethyl)-3-phenyl-1,2,4-oxadiazole

  • MDL number MFCD00512786
  • Cas Number : 1822-94-2
  • Molecular Weight : 194.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart