LIF189457572 Life Chemicals Inc.

2-(cyclopentylsulfanyl)acetic acid

  • MDL number MFCD09944389
  • Cas Number : 52363-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart