LIF189457603 Life Chemicals Inc.

Methyl 3-acetyl-1H-indole-6-carboxylate

  • MDL number MFCD16631783
  • Cas Number : 106896-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart