LIF189457617 Life Chemicals Inc.

5-(bromomethyl)thiophene-2-carbonitrile

  • MDL number MFCD08236857
  • Cas Number : 134135-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart