LIF189457717 Life Chemicals Inc.

2-Amino-5,6-dihydro-4H-cyclopenta[d][1,3]thiazole-4-carboxylic acid

  • MDL number MFCD11909837
  • Cas Number : 1105191-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart