LIF189457725 Life Chemicals Inc.

4-hydroxy-5-methoxypyridine-2-carboxylic acid

  • MDL number MFCD09965920
  • Cas Number : 51727-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart