LIF189457822 Life Chemicals Inc.

methyl 6-hydroxy-1H-indole-2-carboxylate

  • MDL number MFCD11053690
  • Cas Number : 116350-38-0
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart