LIF189457823 Life Chemicals Inc.

6-hydroxy-1H-indole-2-carboxylic acid

  • MDL number MFCD01549019
  • Cas Number : 40047-23-2
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart