LIF189457827 Life Chemicals Inc.

4-hydroxy-1H-indole-2-carboxylic acid

  • MDL number MFCD13178446
  • Cas Number : 80129-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart